J.T.Baker®, Robotic Tips Type HT
Supplier: Avantor
49008-0004EA 0 INR
49008-0004 49008-0104 49008-0105 49009-0005 49009-0105 49010-0105 49008-1105 49009-1005 49009-1105 49010-1005 49010-1105 49008-0003 49010-0003 49010-3105 49010-3005 49008-3105 49009-3105 49009-3005 49008-2003 49009-2005 49009-2105 49010-2003 49010-2105
J.T.Baker®, Robotic Tips Type HT
Pipette Tips
Suitable for use on third party robotic instruments Hamilton Microlab Star and Nimbus. Available in conductive version, with or without filter, and in CE-IVD marked version for clinical diagnostic applications.
  • Manufactured under cleanroom conditions (cleanroom Class 8)
  • State-of-the-art quality control
  • Ideal for high throughput analysis
  • Barcoded trays for automated identification
  • Packed in blister
J.T.Baker® robotic tips type HT are suitable for third party robotic instruments Hamilton Microlab Star, Nimbus, and other robotic instruments usingMore Product Information