Tubes Cryo.S™ 4 and 5 ml
Supplier: Greiner Bio-One
124263_300
479-4150EA 0 INR
479-4150
Tubes Cryo.S™ 4 and 5 ml
Vials
For storage in the freezer or in liquid nitrogen vapor. Suitable for temperatures from −196 to +121 °C.
  • Certified non-cytotoxic and free of detectable endotoxins, DNase/RNase and human DNA
  • With writing area and graduation
PP, with coloured cap, sterile (SAL 10⁻³).
* without writing area