VWR® Eco Premium ULT Freezers
Supplier: VWR Collection
471-1252EA 1434058.5 INR
471-1252 471-1253 471-1304
VWR® Eco Premium ULT Freezers
Freezers Ultra Low Temperature Freezers
VWR® ULT Eco Premium freezers are designed for safe and reliable storage of sensitive samples. They feature ergonomic control and low energy consumption.
  • Maximum 2'' box capacity: 264/352/528
  • Energy star certified
  • Temperatures between −86 and −50 °C
  • Eco Premium 352 and 528: Long warm up time (warm up time from −86 and −50 °C: 360 min)
  • Eco Premium 264: Long warm up time (warm up time from −86 and −50 °C: 280 min)
  • Highly efficient VIP insulation and long lasting rust proof sample chamber
  • Green refrigerants
  • USB and Ethernet interface for data transfer