VWR®, Ultra Low Temperature Freezer, -40 to -86 ℃
Supplier: VWR Collection
471-1274EA 632661 INR
471-1274 471-1275 471-1276 471-1277 471-1282
VWR®, Ultra Low Temperature Freezer, -40 to -86 ℃
Freezers Ultra Low Temperature Freezers
This product line is designed and manufactured for long term storage of various biological products, including viruses, germs, erythrocytes and leucocytes.
  • Special V-I-P (Vacuum Insulation Panel) insulation system reduces the heat gain by 25%
  • High efficiency HC refrigeration system improves the overall efficiency by 45%
  • Microprocessor-controlled system designed for controllable range of -40 to -86 °C for cabinet space with 1 °C increments
  • Large LED display for cabinet temperature, set temperature, ambient temperature, and input voltage
  • Cabinet insulation
More Product Information