Universal freezer, ventilated, NoFrost
Supplier: Liebherr
GN1056
471-0408EA 0 INR
471-0408
Universal freezer, ventilated, NoFrost
Freezers Laboratory / General Purpose Freezers
GN series no-frost laboratory freezer.
  • With cooling accumulators
  • SuperFrost function, temperature alarm
  • Automatic defrosting
  • Interchangeable door hinge
Temperature: −20 °C