Hydrochloric acid concentrate for dilution to 1 mol aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

4657-01EA 0 INR
4657-01
Hydrochloric acid concentrate for dilution to 1 mol aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Hydrochloric acid

(1 equiv.=36,46 g HCl)

Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 0.998-1.002
Normality (by titrimetry) Passes Test
(Makes 1N solution after dilution to 1000 mL)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR