Accessories for Optima™ heated circulating baths
Supplier: GRANT INSTRUMENTS
QR-13 QR-24 QR-LE QR-SE RS14 RS22 RS28 RS38 VR-13 VR-19 VR-24 VR-30 VR-LE VR-SE QR-30
462-8251EA 0 INR
462-8251 462-8253 462-8256 462-8255 462-8263 462-8264 462-8265 462-8266 462-8257 462-8258 462-8133 462-8134 462-8262 462-8261 462-8254
Accessories for Optima™ heated circulating baths
Circulating Thermostats Open Circulator Systems
A range of different stainless steel accessories to use with Optima™ baths.
  • Raised shelves help accommodate different sizes of vessel by altering the effective liquid depth
  • Robust test tube racks are available in a variety of configurations for all sizes of tubes
Ordering information: ST5 and P5 baths can accommodate 1 of the following racks: 462-8251, 462-8252, 462-8253, 462-8254, 462-8255 and 462-8256, the ST12 or P12 and larger baths can accommodate 2 or more of the following racks 462-8257, 462-8258, 462-8133, 462-8134, 462-8261 or 462-8262.