Boiling baths, SBB Aqua Plus series
Supplier: GRANT INSTRUMENTS
SBBAQUA12PLUS SBBAQUA26PLUS SBBAQUA5PLUS
462-0166EA 0 INR
462-0166 462-0170 462-0171
Boiling baths, SBB Aqua Plus series
Baths Boiling Baths
Boiling baths are designed for continuous operation with water above 90 °C. They have a stainless steel tank in an outer case and analogue control by means of a simple dial for straightforward temperature settings. Baths have two fixed over-temperature safety cut-outs which can be reset by the user.
  • Energy regulator can be adjusted to ensure a steady boiling action, avoiding excessive steam or spitting
  • Constant level device maintains the required liquid level, avoiding the need to monitor and top up the bath
  • Heater and temperature sensors are mounted at
More Product Information