VWR®, Accessories for Refrigerated Circulators
Supplier: VWR Collection
461-0417EA 47936.7 INR
461-0417
VWR®, Accessories for Refrigerated Circulators
Circulating Thermostats Open Circulator Systems
For use with VWR® circulators with a programmable or digital controller when direct digital control is not required. Connects to the RS232 port and provides 10 mV/degree analogue signal for external monitoring or control.