VWR® ARISTAR®, Tellurium Standard Solutions for ICP-MS
Supplier: VWR Chemicals
85634.180EA 13034.25 INR
85634.180 457461A
VWR® ARISTAR®, Tellurium Standard Solutions for ICP-MS
Standards Cationic Standards Tellurium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0.5% (typical)
  • Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities