VWR® ARISTAR®, Beryllium Standard Solution, 10000 mg/L Be in dil. Nitric Acid, Standard for ICP
Supplier: VWR Chemicals
455092UEA 45704.51 INR
455092U
VWR® ARISTAR®, Beryllium Standard Solution, 10000 mg/L Be in dil. Nitric Acid, Standard for ICP
Standards Cationic Standards Beryllium Standards
BeO.(Be(OAc)₂)₃) in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.