Hotplates, C-MAG HP series
Supplier: IKA JANKE KUNKEL
3581800 3582002 3581600 3581602 3581603 3581802 3581803 3582000 3582003
453-0087EA 0 INR
453-0087 553-0002 453-0070 553-0000 453-0071 553-0001 453-0072 453-0073 453-0074
Hotplates, C-MAG HP series
Heaters Hotplates
Compact hotplates with a chemically resistant ceramic top. The elevated control panel minimises contact with spills and enables easy reading and adjustment of the temperature. Units feature a digital error code display and 'hot' top indicator to warn user that the surface is hot.
  • C-MAG HP 7 and C-MAG HP 10 can be used with e.g. ETS-D5 contact thermometer, enabling precise temperature control
  • Fixed safety circuit shuts off heater if temperature rises above 550 °C
  • Microprocessor control sends feedback to the hotplate to maintain set temperature
  • Digital
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable