VWR®, Microplate Shaker, High Speed
Supplier: VWR Collection
14216-214 13500-890
444-7016EA 1004139.36 INR
444-7016 VWRU14216-214 VWRU13500-890
VWR®, Microplate Shaker, High Speed
Shakers Microplate Shakers
Ideal for molecular biology applications, mechanical and chemical cell lysis, mixing tissue samples, mixing cytogenetic suspensions, and vortexing cell suspensions. Shaker is also ideal for emulsification of bi-polar viscous fluids and for mixing fluids in small diameter tubes. Specifically designed to shake and/or vortex microplates in timed or continuous modes. Shaker features programmable timed mode from 1 s to 160 hours. Speed and time are easily adjusted with the up/down arrows on the front panel.
  • Holds up to 6 microplates without stacking, up to a maximum of 48 microplates
More Product Information