VWR® Advanced 700, Hotplates
Supplier: VWR Collection
444-2863EA 0 INR
444-2863 444-2865 444-2867 444-2869
VWR® Advanced 700, Hotplates
Heaters Hotplates
VWR® 700 series advanced hotplates deliver accurate and repeatable results. Low profile, space saving design takes up less space in the lab. Spill resistant housing channels fluids away from internal components. Front panel features easy to use controls, which allow users to dial-in adjustments.
  • LED display and microprocessor control
  • Over-temperature protection
  • On-board memory recalls last set temperature
  • Ceramic or aluminium top plate
  • Hot warning when plate is in use
Integral support rod holder with locking knob accepts the optional support rod andMore Product Information