Rocking platform shakers, Duomax 1030
Supplier: Heidolph Instruments GmbH & Co.KG
543-32205-06 543-32205-02
444-1236EA 0 INR
444-1236 444-1479
Rocking platform shakers, Duomax 1030
Shakers Rotating/Rocking Shakers
Platform shaker with rocking motion, designed for homogeneous mixing, ideal for flasks, media bottles and large vessels. Units have analogue dial controls for rocking frequency and time.
  • Modular system can be combined with the incubator 1000 and a low or high hood
  • Timer function with audible signal allows a process time up to 120 min
  • Choice of units with tilt fixed at 5° or 10° angle, electronic speed control
  • Ideal for use under controlled conditions, can be used at 0 to 50 °C (80% relative humidity), in an incubator, cold room or CO₂ incubator