VWR®, Multi-position Magnetic Stirrer
Supplier: VWR Collection
444-1081EA 93788 INR
444-1081
VWR®, Multi-position Magnetic Stirrer
Stirrers Magnetic Hotplate Stirrers, Multi-position
Designed for synchronous heating and stirring, this robust multi-position magnetic hotplate stirrer accommodates 15×400 ml flasks. Units have a metal housing with an aluminium plate and can be operated with the Labs-connect App via Wifi/USB controls. Three separate LED displays show temperature, speed and time set. The motorless magnetic coil technology ensures noiseless and consistent stirring at all positions and the SPC-mode provides a low self-warming on the plate surface.
  • Homogeneous temperature distribution across the heating plate
  • Hot top warning
More Product Information