Magnetic hotplate stirrers, RH basic and RH digital
Supplier: IKA JANKE KUNKEL
5019800 5019802
442-0989EA 0 INR
442-0989 442-0990
Magnetic hotplate stirrers, RH basic and RH digital
Stirrers Magnetic Hotplate Stirrers
Magnetic hotplate stirrers with stainless steel heating plate, white units have an enamelled plate. All models are easy to operate and have adjustable safety temperature from 50 to 370 °C. High heat output of 600 watts and steel-copper compound hot plate ensures rapid heating.
  • RH basic includes M 10 thread for a support rod
  • Soft-start of the stirring motor for both models
  • Strong magnetic field and wide speed range for volumes up to 15 litres
  • RH digital has digital display of temperature and speed
  • Error code display and touch keypad (RH digital)