Oil bath attachment for IKAMAG® and IKATHERM®
Supplier: IKA JANKE KUNKEL
2829500 2829400
442-0582EA 0 INR
442-0582 442-0581
Oil bath attachment for IKAMAG® and IKATHERM®
Baths Oil Baths
The oil bath attachment H29 and H30 can be used as oil baths together with an IKAMAG® magnetic stirrer with heating or with an IKATHERM® heating plate having a diameter of 135 mm.
  • Easy cleaning and hollow cylindrical
  • Made of aluminum cast
  • Safety grip protects from bath attachment burns
  • Positioning border prevents sliding on the heating plate
Order Now
SPECIFICATIONS
Environmentally Preferable