Sterilised filter bags, Whirl-Pak®
Supplier: NASCO FORT ATKINSON
B01318WA B01348WA
432-0169EA 0 INR
432-0169 432-0170
Sterilised filter bags, Whirl-Pak®
Bags Blender Bags
Sterilised post-manufacturing and specially designed for use with homogeniser blenders. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • 330 micron filter layer collecting, blending, pipetting
  • Convenient write-on strip for easy labelling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring and closing
  • Opening pull tabs: For easy opening, non slip textured Tyvek
  • Continuous
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable