Dispersing elements for homogenisers
Supplier: IKA JANKE KUNKEL
2348000 2404000 2466900 8005100 8006300 8003500 8004100 8003200 3304000 1684300 4446900 8039500 4446500 4447500 4447300 4446700 4447100
431-2455EA 0 INR
431-2455 431-2461 431-2458 431-2581 431-2582 431-0256 431-0257 431-0255 442-0300 431-0249 431-0253 431-0258 431-0251 431-0223 431-0222 431-0252 431-0254
Dispersing elements for homogenisers
Homogenisers Dispersing Tools
Type N: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by all methods, max. temperature 180 °C.
Type NK: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by chemical methods, max. temperature 120 °C.
Type KV: Materials in contact with medium are FFPM/SIC and stainless steel AISI 316L, suitable for vacuum down to 1 mbar, pressure up to 6 bar and for solvents, can be sterilised by chemical methods, max.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable