Zinc acetate dihydrate 98.0-102.0%, Technical Grade, J.T.Baker®

Supplier: Avantor

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

4304-REA 0 INR
4304-R
Zinc acetate dihydrate 98.0-102.0%, Technical Grade, J.T.Baker®
Zinc acetate dihydrate
Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Melting Pt: 237
MDL Number: MFCD00066961
CAS Number: 5970-45-6
UN: 3077
ADR: 9 III
Merck Index: 12,10256

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 98.0 - 102.0 %
Assay ((CH₃COO)₂Zn·2H₂O) 98.0 - 102.0 %
Insoluble Matter ≤0.01 %
pH of 5% Solution at 25°C 6.0 - 7.0
Lead (Pb) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR