Starch, soluble Potato, powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor

Synonyms: Starch, Vitex® iodine indicator, Amylodextrin

4006-01EA 78747.33 INR
4006-01 4006-05 4006-04
Starch, soluble Potato, powder, BAKER ANALYZED® ACS, J.T.Baker®
Starch, soluble

For iodometry.


  • Lot analysis on label

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 5.0 - 7.0
For Laboratory,Research,or Manufacturing Use ≤0.4%
Solubility Passes Test
pH of 2% Solution at 25°C 5.0 - 7.0
Residue after Ignition ≤0.4 %
Sensitivity Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR