Sodium fluoride 98,0-102,0 Dry_basis, powder USP, J.T.Baker®

Supplier: Avantor
Danger

3689-08EA 226116.63 INR
3689-08
Sodium fluoride 98,0-102,0 Dry_basis, powder USP, J.T.Baker®
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700 °C (1013 hPa)
Melting Pt: 993 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1 III
Merck Index: 13,08691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product Passes Test
Meets U.S.P Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤1.0%
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity or Alkalinity Passes Test
USP - Loss on Drying ≤1.0 %
USP - Fluosilicate (volume of 0.10N NaOH required) ≤1.5 ml
USP - Chloride (Cl) ≤0.012 %
USP - Assay (NaF) (dried basis) 98.0 - 102.0 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR