Soda lime, BAKER ANALYZED® ACS (4-8 Mesh) Indicator, J.T.Baker®

Supplier: Avantor
Warning

3447-01EA 17045.21 INR
3447-01
Soda lime, BAKER ANALYZED® ACS (4-8 Mesh) Indicator, J.T.Baker®
Soda lime
MDL Number: MFCD00134094
CAS Number: 8006-28-8
UN: 1907
ADR: 8 III
Merck Index: 13,08640

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Loss on Drying at 200°C ≤7 %
ACS - Carbon Dioxide Absorption Capacity ≥19 %
ACS - Presence of Indicator Passes Test
ACS - Fines ≤1 %
Mesh - On U.S. No. 4 Sieve ≤10 %
Mesh - Thru U.S. No. 8 Sieve ≤25 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR