Silica gel, BAKER ANALYZED® 60 - 200 mesh Chromatography_column, J.T.Baker®

Supplier: Avantor

3414-01EA 58368.78 INR
3414-01 3414-05
Silica gel, BAKER ANALYZED® 60 - 200 mesh Chromatography_column, J.T.Baker®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use Passes Test
Toluene Absorption Passes Test
Loss on Ignition ≤ 10.0 %
Alcohol-Ether Soluble Substances ≤ 0.10 %
Bulk Density (g/cc), For Information Only = 15 %
Mesh - On U.S. No. 60 Sieve ≤ 15 %
Mesh - Thru U.S. No. 200 Sieve ≤ 15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR