Methyl blue stain_microbiology (C.I. 42780)

Supplier: VWR Chemicals

Synonyms: Aniline blue sodium salt, [[4-[bis[4-[(sulphophenyl)amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]amino]-benzenesulphonic acid sodium salt (1:2), Poirriers blue, Cotton blue, Water blue, C.I. 42780, Aniline blue, water soluble, Disodium [[4-[bis[4-[(sulphonatophenyl)amino]phenyl]methylene]cyclohexa-2,5-dien-1-ylidene]amino]benzenesulphonate, Acid Blue 93

34015.182EA 0 INR
34015.182
Methyl blue stain_microbiology (C.I. 42780)
Methyl blue
Formula: C₃₇H₂₇N₃Na₂O₉S₃
MW: 799.81 g/mol
MDL Number: MFCD00058509
CAS Number: 28983-56-4
Merck Index: 13,06055

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR