VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
331-0383EA 0 INR
331-0383 331-0384 331-0385 331-0386 331-0387 331-0388 331-0389 331-0390 331-0391 331-0392 331-0393 331-0394 331-0395 331-0396 331-0397 331-0398 331-0399 331-0400 331-0401 331-0402 331-0403 331-0404 331-0405 331-0406 331-0407 331-0408 331-0409
VWR® Water Sampling Bottles, with Tamper Evident Cap
Bottles Water Sampling Bottles
HDPE water sampling collection bottle.
  • Bottle made with HDPE resin
  • Natural bottle colour, with red cap
  • Supplied sterile
  • Sodium thiosulphate in powder form
  • Product labelled with unique barcode and sample collection write-on for traceability
  • Tamper-evident cap options with 33 mm pull type, 45 and 63 mm threaded cap
Wide neck design allows for easy sample collection. Product labelled with dosage and write-on details. These bottles are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the waterMore Product Information