Ammonium acetate 97,0, crystals, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: Acetic acid ammonium salt

3272-20EA 63003 INR
3272-20 3272-04 MACR3272-06
Ammonium acetate 97,0, crystals, AR® ACS, Macron Fine Chemicals™
Ammonium acetate
Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Density: 1.17 g/cm³ (20 °C)
Flash Pt: 136
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use ≥97%
ACS - Assay (CH₃COONH₄) ≥97 %
ACS - Appearance (colorless crystals) Passes Test
ACS - Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Nitrate (NO₃) ≤0.001 %
ACS - pH of 5% Solution at 25°C 6.7 - 7.3
ACS - Residue after Ignition ≤0.01 %
ACS - Sulfate (SO₄) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR