di-Sodium L(+)-tartrate dihydrate 99, GPR RECTAPUR®

Supplier: VWR Chemicals

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

28174.260EA 0 INR
28174.260
di-Sodium L(+)-tartrate dihydrate 99, GPR RECTAPUR®
di-Sodium L(+)-tartrate dihydrate
Formula: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
Melting Pt: 57
Density: 1.818 g/cm³ (20 °C)
MDL Number: MFCD00150035
CAS Number: 6106-24-7
EINECS: 212-773-3
Merck Index: 12,08833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39a75038f4ff5e6c234267b15540823ae361bed4&vid=89289572c48b0c6415f8f11e916a51f66e129474&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 99 %
Loss on drying (150°C) Max. 17.0 %
C2O4 (Oxalate) Max. 0.05 %
Cl (Chloride) Max. 50 ppm
As (Arsenic) Max. 2 ppm
Cu (Copper) Max. 10 ppm
Fe (Iron) Max. 100 ppm
Pb (Lead) Max. 10 ppm
Zn (Zinc) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR