Pyridine, anhydrous 99,7, AnalaR NORMAPUR® Reag. Ph. Eur., ACS, ISO zA, Karl_Fischer

Supplier: VWR Chemicals
Danger

Synonyms: Azabenzene, Azine

27198.298EA 0 INR
27198.298
Pyridine, anhydrous 99,7, AnalaR NORMAPUR® Reag. Ph. Eur., ACS, ISO zA, Karl_Fischer
Pyridine
Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colourless liquid
Assay (GC) Min. 99.7 %
Acidity or alkalinity Max. 0.200 meq/g
Water Max. 0.0200 %
Evaporation residue Max. 0.0020 %
Reducing substances Max. 0.0005 %
Piperidine Max. 0.0100 %
2-Methylpyridine Max. 0.20 %
NH4 (Ammonium) Max. 0.0010 %
Cl (Chloride) Max. 0.0005 %
SO4 (Sulphate) Max. 0.0005 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.05 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.01 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR