L(+)-Potassium hydrogen tartrate 99,0-101,0 Ph. Eur.

Supplier: VWR Chemicals

Synonyms: Potassium hydrogen L-tartrate, Potassium acid tartrate, Potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Potassium Bitartrate, (R,R)-(+)-Tartaric acid potassium salt, L(+)-Tartaric acid potassium salt

27057.361EA 0 INR
27057.361
L(+)-Potassium hydrogen tartrate 99,0-101,0 Ph. Eur.
L(+)-Potassium hydrogen tartrate
Formula: KOOC(CHOH)₂COOH
MW: 188.18 g/mol
Melting Pt: 250
Density: 1.943 g/cm³ (20 °C)
MDL Number: MFCD00065392
CAS Number: 868-14-4
EINECS: 212-769-1
Merck Index: 13,07697
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=355f72f319f5589f12d60c6dfffecdec31cee083&vid=4a750ad5d4a91fd4c2885832ee80b7b64cdcc42a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on dried substance) 99.0 - 101.0 %
Appearance Conforms (see CoA/CoS)
Identification B Passes test
Identification C Passes test
Identification D Passes test
Spec. opt. rotation (dried substance) 8 - 9.2 °
Oxalic acid Max. 500 ppm
Cl (Chloride) Max. 500 ppm
SO4 (Sulphate) Max. 500 ppm
Heavy metals (as Pb) Max. 10 ppm
Loss on drying (105°C) Max. 0.5 %
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR