Potassium sulphate 99,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA

Supplier: VWR Chemicals

26997.293EA 10054.99 INR
26997.293
Potassium sulphate 99,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Assay Min. 99.0 %
pH (25°C; 5 %) 5.5 - 7.5
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 100 ppm
Total N (Nitrogen) Max. 5 ppm
Cl (Chloride) Max. 5 ppm
As (Arsenic) Max. 2 ppm
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 20 ppm
Na (Sodium) Max. 0.02 %
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR