2-Phenoxyethanol 99,0-100,5 Ph. Eur.

Supplier: VWR Chemicals
Warning

Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether

26244.290EA 15244.91 INR
26244.290
2-Phenoxyethanol 99,0-100,5 Ph. Eur.
2-Phenoxyethanol
Formula: C₆H₅OCH₂CH₂OH
MW: 138.17 g/mol
Boiling Pt: 245
Melting Pt: 12 °C
Density: 1.1094 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002857
CAS Number: 122-99-6
EINECS: 204-589-7
Merck Index: 12,07410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27cc4d39b806528798164961c23a1285d41d6a9&vid=a46b721cc69101d0c50dc49d9687445b395a3766&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 100.5 %
Appearance C'less slightly visc.liq.
Identification (IR) Passes test
Relative density 1.105 - 1.11
Unspecified impurities (for each) Max. 0.10 %
Total impurities Max. 0.3 %
Reporting treshold Max. 0.05 %
Residual solvents Passes test
As (Arsenic) ≤ 1 ppm
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 1 ppm
Ni (Nickel) ≤ 1 ppm
Pb (Lead) ≤ 1 ppm
V (Vanadium) ≤ 1 ppm
Conforms to Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR