Clamps, universal
Supplier: USBECK LABORGERATE
2505
241-7311EA 0 INR
241-7311
Clamps, universal
Clamps General Purpose Clamps
The fingers of the clamps have cork inserts to protect the clamped items.
  • Without bosshead for fixing to stand
  • With T-screws for rapid easy clamping
Order Now
SPECIFICATIONS
Environmentally Preferable