Stand bases
Supplier: USBECK LABORGERATE
2013
241-7175EA 0 INR
241-7175
Stand bases
Retort Stands
Stainless steel. Firm, non slip stand for a wide variety of uses. With an M10 threaded hole on face side and adjustable feet for uneven surfaces.
  • Rubber feet for a firm footing
  • Smooth surfaces for easy, fast cleaning
  • Robust
DIN 12892/55
Certifications: DIN 12892/55