VWR®, Laboratory Stands
Supplier: VWR Collection
241-0090EA 0 INR
241-0090 241-0091 241-0092 241-0093 241-0094 241-0095 241-0099 241-0100 241-0101
VWR®, Laboratory Stands
Retort Stands
Tripod stands (A): Cast iron tripod stand, painted with hammer finish, steel support rod, nickel plated.
  • Threaded hole
  • Support rod (three lengths available with different diameters)
  • Robust, secure stand
- Threaded hole
- Support rod (three lengths available with different diameters)
- Robust, corrosion resistant
Support stands (B): Cast iron stand base, painted with hammer finish, steel support rod.
- Eccentrically-attached support rod
- Firm, non slip stand
- Available in three different sizes
- Robust, corrosion resistant
Support stands (C):More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable