Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

23840.292EA 0 INR
23840.292 23840.326 23840.460
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
Petroleum spirit
Boiling Pt: 60…80 °C (1013 hPa)
Density: 0.67 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Order Now

Specification Test Results

Acidity Max. 0.0002 meq/g
Bromine value Max. 3
Colouration Max. 10 APHA
Substances coloured by H2SO4 Max. 150 APHA
Boiling range (60-80°C) Min. 90 %
Evaporation residue Max. 10 ppm
Sulphur compounds (as CS2) Max. 3 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.15 ppm
Ba (Barium) Max. 0.08 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.3 ppm
Mg (Magnesium) Max. 0.08 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.75 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.08 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.03 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR