Double spatulas
Supplier: BOCHEM
31045
231-0042EA 0 INR
231-0042
Double spatulas
Spatulas
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • Round handle, one flat and one rounded end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance