Peristaltic pump drives
Supplier: Heidolph Instruments GmbH & Co.KG
523-52013-00 523-52060-00 523-52010-02 523-52060-02 523-52013-02 523-52060-03 523-52013-03
224-0756EA 0 INR
224-0756 224-1355 224-1459 224-1438 224-1435 HEID523-52060-03 HEID523-52013-03
Peristaltic pump drives
Pumps Pump Drives Peristaltic Pump Drives
Models PD 5201 and PD 5206 for advanced requirements. Peristaltic pumps are ideal for transporting and metering aggressive, abrasive, or corrosive solutions, and also for sterile media under low pressure. The medium to be transported only comes into contact with the tubing, so there is no contamination. Suitable for transporting highly viscous media up to 10000 mPas. The pumps are self-priming without seals or valves. Can be used in the food industry for filtering and separating, for water treatment, in biotechnology and routine laboratory applications. Media can be transported underMore Product Information