VWR®, Conical Stoppers, Cork
Supplier: VWR Collection
217-1011EA 0 INR
217-1011 217-1012 217-1013 217-1014 217-1015 217-1016 217-1017 217-1018 217-1019 217-1020 217-1021 217-1022 217-1023 217-1024 217-1025 217-0897 217-0898
VWR®, Conical Stoppers, Cork
Stoppers Cork Stoppers
Natural cork, A1 quality.
  • Lightweight, tough
  • Easy to drill holes into
  • Very economical
Order Now
SPECIFICATIONS
Environmentally Preferable