Specimen jars with ground-in knobbed lid, DURAN®
Supplier: DWK LIFE SCIENCES
242090207 242090901 242091109 242091606 242091709 242092405 242092602 242092808 242093804 242093907 242094903 242095008 242095702 242095908
216-3331EA 0 INR
216-3331 216-3332 216-3333 216-3340 216-3334 216-0285 216-3335 216-3339 SCOT242093804 216-3336 SCOT242094903 SCOT242095008 216-3337 216-3338
Specimen jars with ground-in knobbed lid, DURAN®
Jars Specimen Jars
DURAN®, borosilicate glass 3.3, clear, with ground joint. Low profile.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Order Now
SPECIFICATIONS
Environmentally Preferable