VWR® Ice Buckets, Rectangular, with Lid
Supplier: VWR Collection
216-1510EA 0 INR
216-1510 216-1513 216-1514 216-1515 216-1516 216-1517 216-1518 216-1519
VWR® Ice Buckets, Rectangular, with Lid
Ice Containers
Polyurethane foam.
  • Resistant to temperatures from –76 to +70 °C
  • Extremely robust, leakproof, stackable, lightweight
  • Available in two sizes and four colours
  • Stacking option for saving space on working bench
  • Pouring lip design, for easy draining of liquid
  • With lid to reduce the evaporation level of dry ice, water ice with salt slurry
Ideal for storing samples, these buckets maintain hot or cold temperatures of bottles, test tubes, solutions, and cultures. Suitable for use with dry ice, water ice, salt solution. For freezer storage only.
Order Now
SPECIFICATIONS
Environmentally Preferable