VWR® Erlenmeyer Flasks, Conical, with Interchangable Joint
Supplier: VWR Collection
214-0312EA 0 INR
214-0312
VWR® Erlenmeyer Flasks, Conical, with Interchangable Joint
Flasks Erlenmeyer Flasks
Borosilicate glass, grade 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
The good value alternative for a wide range of laboratory applications. Volume graduations and labelling area.
Certifications: DIN 12387