Flasks, round bottom, narrow or wide neck, with flared rim, DURAN®
Supplier: DWK LIFE SCIENCES
217211706 217212402 217213604 217215408 217216404 217216807 217217109 217217306 217217709 217218602 217219101 217412407 217414408 217415404 217416306 217416409 217417105 217417302 217417602 217418607 217419106 217214403 217218705 217413609
214-2105EA 0 INR
214-2105 214-2110 214-2115 214-2125 214-2130 214-2135 214-0146 214-2145 214-2150 214-2155 214-2160 214-2210 214-2220 214-2225 214-2231 214-2230 214-2240 214-2245 214-2251 214-2255 214-2260 214-2120 214-0427 214-2215
Flasks, round bottom, narrow or wide neck, with flared rim, DURAN®
Flasks Round Bottom Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area. Flasks with neck external diameter larger than 65 mm have reinforced rims.
*Non-DIN ISO size
DIN EN ISO 24450 (wide mouth)
Certifications: DIN ISO 1773 (narrow neck)
DIN EN ISO 24450 (wide neck)