Culture tubes, disposable, KIMBLE®
Supplier: KIMBLEKONTES
73500-1275
212-0625EA 0 INR
212-0625
Culture tubes, disposable, KIMBLE®
Tubes Culture Tubes
Manufactured from 51 expansion borosilicate glass.
  • Offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
  • The 6×50 mm size is often referred to as a 'Durham tube'
These premium quality tubes feature sturdy, uniform bottoms and consistent lengths. The 10×75 and 12×75 mm sizesMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable