Aluminium oxide activated basic Chromatography

Supplier: VWR Chemicals

21012.297EA 10567 INR
21012.297
Aluminium oxide activated basic Chromatography
Aluminium oxide activated basic
Formula: Al₂O₃
MW: 101.96 g/mol
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Specification Test Results

Identification Passes test
Particle size ( 48 µm) Max. 20 %
Particle size ( 200 µm) Max. 1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR