Sulphuric acid 95% 95, AnalaR NORMAPUR® zA, Trace_analysis_Cd_Hg_Pb

Supplier: VWR Chemicals
Danger

20704.292EA 0 INR
20704.292 20704.320
Sulphuric acid 95% 95, AnalaR NORMAPUR® zA, Trace_analysis_Cd_Hg_Pb
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 10.38 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
REACH: 01-2119458838-20

Order Now

Specification Test Results

Assay Min. 95.0 %
Colouration Max. 10 APHA
Ignition residue Max. 5 ppm
Substances reducing KMnO4 (as SO2) Max. 5 ppm
Cl (Chloride) Max. 0.2 ppm
NH4 (Ammonium) Max. 2 ppm
NO3 + NO2 (as NO3) Max. 0.1 ppm
As (Arsenic) Max. 0.01 ppm
Cd (Cadmium) Max. 0.005 ppm
Cu (Copper) Max. 0.05 ppm
Fe (Iron) Max. 0.2 ppm
Hg (Mercury) Max. 0.005 ppm
Ni (Nickel) Max. 0.05 ppm
Pb (Lead) Max. 0.005 ppm
Zn (Zinc) Max. 0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR