Nitric acid 68%, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Nitric acid solution

20406.320EA 2900 INR
20406.320
Nitric acid 68%, TECHNICAL
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 (5.1) II
Merck Index: 13,06608

Order Now

Specification Test Results

Assay 68 - 70 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR