Acetonitrile 99,9, HiPerSolv CHROMANORM®, isocratic HPLC

Supplier: VWR Chemicals
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

Filter 0,2 um_Nitrogen_filled

20048.320EA 0 INR
20048.320 20048.290 20048.360
Acetonitrile 99,9, HiPerSolv CHROMANORM®, isocratic HPLC
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Acidity Max. 0.0003 meq/g
Evaporation residue Max. 5 ppm
Absorbance (200 nm) Max. 0.1
Absorbance (210 nm) Max. 0.1
Absorbance (220 nm) Max. 0.05
Absorbance (230 nm) Max. 0.01
Absorbance (250 nm) Max. 0.005
Transmittance (200 nm) Min. 80 %
Transmittance (210 nm) Min. 85 %
Transmittance (220 nm) Min. 90 %
Transmittance (230 nm) Min. 98 %
Transmittance (250 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR