Ammonium iron (III) citrate, red brown, powder FCC, J.T.Baker®

Supplier: Avantor

Synonyms: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

1980-01EA 0 INR
1980-01
Ammonium iron (III) citrate, red brown, powder FCC, J.T.Baker®
Ammonium iron (III) citrate
Formula: C₆H₅+4y)Fe(x)N(y)O₇
Density: 1.8 g/cm³ (20 °C)
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6

Order Now

Specification Test Results

Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical 16.5 - 18.5 %
FCC - Assay (as Fe) 16.5 - 18.5 %
Identification Passes Test
Ferric Citrate Passes Test
Lead (Pb) ≤2 mg/kg
Mercury (Hg) ≤1 mg/kg
Oxalate (C₂O₄) Passes Test
Sulfate (SO₄) ≤0.3 %
Tartrate Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR